ENA282547990 Enamine

N'-propyl-2-(propylamino)butanediamide

  • MDL number MFCD15209657
  • Cas Number : 1251924-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60972
Availability details for
Quantity
  • Availability
  • Add to Cart