ENA282547997 Enamine

[2-(morpholin-4-yl)ethyl]urea hydrochloride

  • MDL number MFCD15209664
  • Cas Number : 1235440-75-1
  • Molecular Weight : 209.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61042
Availability details for
Quantity
  • Availability
  • Add to Cart