ENA282547998 Enamine

4-{[(cyclopropylmethyl)amino]methyl}phenol hydrochloride

  • MDL number MFCD15209665
  • Cas Number : 1221722-75-3
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61045
Availability details for
Quantity
  • Availability
  • Add to Cart