ENA282548001 Enamine

2-(aminomethyl)phenol hydrochloride

  • MDL number MFCD15209668
  • Cas Number : 61626-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61062
Availability details for
Quantity
  • Availability
  • Add to Cart