ENA282548006 Enamine

1-[1-(2-bromophenyl)-1H-1,2,3-triazol-4-yl]methanamine hydrochloride

  • MDL number MFCD15209673
  • Cas Number : 1221725-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61119
Availability details for
Quantity
  • Availability
  • Add to Cart