ENA282548009 Enamine

4-(2-aminoethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one hydrochloride

  • MDL number MFCD15209676
  • Cas Number : 1235439-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212...214
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61131

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61131
Availability details for
Quantity
  • Availability
  • Add to Cart