ENA282548017 Enamine

1-methyl-1,2,3,4-tetrahydroquinoline-6,8-dicarbaldehyde

  • MDL number MFCD15209685
  • Cas Number : 68032-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61196
Availability details for
Quantity
  • Availability
  • Add to Cart