ENA282548020 Enamine

3-methyl-N-(piperidin-4-yl)-1,2-oxazole-5-carboxamide hydrochloride

  • MDL number MFCD15209688
  • Cas Number : 1235440-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61216
Availability details for
Quantity
  • Availability
  • Add to Cart