ENA282548021 Enamine

2-chloro-N-[2-(dimethylamino)ethyl]acetamide hydrochloride

  • MDL number MFCD15209689
  • Cas Number : 92384-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61219
Availability details for
Quantity
  • Availability
  • Add to Cart