ENA282548026 Enamine

2-(5-amino-1H-indol-1-yl)-N,N-diethylacetamide hydrochloride

  • MDL number MFCD15209694
  • Cas Number : 1235441-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61237
Availability details for
Quantity
  • Availability
  • Add to Cart