ENA282548032 Enamine

2,2,2-trifluoroethyl piperidine-1-carboxylate

  • MDL number MFCD15209700
  • Cas Number : 1092074-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61303

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61303
Availability details for
Quantity
  • Availability
  • Add to Cart