ENA282548038 Enamine

2-[(2-methoxyethyl)amino]acetamide hydrochloride

  • MDL number MFCD15209706
  • Cas Number : 1235440-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61352

Safety Information

signalword Warning
pcodes P261,P264,P271,P272,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P333+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-61352
pictograms GHS07
hstatements H315,H317,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart