ENA282548040 Enamine

2,2,2-trifluoroethyl N-(3-acetylphenyl)carbamate

  • MDL number MFCD15209708
  • Cas Number : 1221723-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61367
Availability details for
Quantity
  • Availability
  • Add to Cart