ENA282548041 Enamine

3-amino-2-(2-chlorophenyl)-1,1,1-trifluoropropan-2-ol hydrochloride

  • MDL number MFCD15209710
  • Cas Number : 1221726-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61387
Availability details for
Quantity
  • Availability
  • Add to Cart