ENA282548043 Enamine

3-methyl-N-(piperidin-4-yl)furan-2-carboxamide hydrochloride

  • MDL number MFCD15209712
  • Cas Number : 1221725-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61401
Availability details for
Quantity
  • Availability
  • Add to Cart