ENA282548047 Enamine

4-phenyl-1-(prop-2-yn-1-yl)-1H-imidazol-5-amine

  • MDL number MFCD15209716
  • Cas Number : 1221724-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61422

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61422
Availability details for
Quantity
  • Availability
  • Add to Cart