ENA282548048 Enamine

2-(5-amino-4-phenyl-1H-imidazol-1-yl)acetamide

  • MDL number MFCD15209717
  • Cas Number : 1235438-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61423
Availability details for
Quantity
  • Availability
  • Add to Cart