ENA282548050 Enamine

2-amino-N-({8-methyl-8-azabicyclo[3.2.1]octan-3-yl}methyl)acetamide dihydrochloride

  • MDL number MFCD19442666
  • Cas Number : 1235438-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61443
Availability details for
Quantity
  • Availability
  • Add to Cart