ENA282548084 Enamine

[2-(pyridin-2-yl)-1,3-thiazol-4-yl]methanol

  • MDL number MFCD11505651
  • Cas Number : 173838-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60482
Availability details for
Quantity
  • Availability
  • Add to Cart