ENA282548089 Enamine

5-(5-methylthiophen-2-yl)thiophene-2-carboxylic acid

  • MDL number MFCD07375985
  • Cas Number : 30979-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60488
Availability details for
Quantity
  • Availability
  • Add to Cart