ENA282548091 Enamine

1-methyl-3-(2-methylpropyl)-1H-pyrazol-5-amine

  • MDL number MFCD11209667
  • Cas Number : 118430-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60491
Availability details for
Quantity
  • Availability
  • Add to Cart