ENA282548093 Enamine

N'-hydroxy-6-methoxypyridine-3-carboximidamide

  • Cas Number : 478490-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60493
Availability details for
Quantity
  • Availability
  • Add to Cart