ENA282548098 Enamine

2-[(4-methoxyphenyl)methoxy]acetonitrile

  • MDL number MFCD11147566
  • Cas Number : 1020944-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60500

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60500
Availability details for
Quantity
  • Availability
  • Add to Cart