ENA282548103 Enamine

1-[(pyrrolidin-2-yl)methyl]-2,3-dihydro-1H-indole

  • MDL number MFCD11155240
  • Cas Number : 1021015-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60506
Availability details for
Quantity
  • Availability
  • Add to Cart