ENA282548104 Enamine

1-(prop-2-yn-1-yl)-1H-pyrrole

  • MDL number MFCD12090928
  • Cas Number : 19016-98-9
  • Molecular Weight : 105.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60507
Availability details for
Quantity
  • Availability
  • Add to Cart