ENA282548106 Enamine

6-(2-chlorophenyl)-3-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09738837
  • Cas Number : 954263-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55242
Availability details for
Quantity
  • Availability
  • Add to Cart