ENA282548112 Enamine

6-(2-methoxyphenyl)-3-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09737322
  • Cas Number : 954253-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55248
Availability details for
Quantity
  • Availability
  • Add to Cart