ENA282548113 Enamine

pyridine-2-carbonyl chloride hydrochloride

  • MDL number MFCD00060193
  • Cas Number : 39901-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-55340

Safety Information

signalword Danger
pcodes P234,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P390,P405,P406,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-55340
pictograms GHS05
hstatements H290,H314
Availability details for
Quantity
  • Availability
  • Add to Cart