ENA282548116 Enamine

5-(3-bromo-5-fluorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD11619414
  • Cas Number : 1094640-20-6
  • Molecular Weight : 274.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56152
Availability details for
Quantity
  • Availability
  • Add to Cart