ENA282548117 Enamine

7-methoxy-3,4-dihydro-2H-1-benzopyran-3-carboxylic acid

  • MDL number MFCD00598395
  • Cas Number : 3187-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60549

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60549
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart