ENA282548122 Enamine

1-[(3-bromophenyl)methyl]-1H-1,2,4-triazol-3-amine

  • MDL number MFCD12766718
  • Cas Number : 1183190-06-8
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60554
Availability details for
Quantity
  • Availability
  • Add to Cart