ENA282548124 Enamine

1-(2-methylpropyl)-1H-imidazol-2-amine

  • MDL number MFCD12164605
  • Cas Number : 1184017-89-7
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60556
Availability details for
Quantity
  • Availability
  • Add to Cart