ENA282548127 Enamine

1-(2-methoxyphenyl)butane-1,3-dione

  • MDL number MFCD07772934
  • Cas Number : 56290-53-0
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60559
Availability details for
Quantity
  • Availability
  • Add to Cart