ENA282548131 Enamine

(2-methylphenyl)thiourea

  • MDL number MFCD00041162
  • Cas Number : 614-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28847

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-28847
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart