ENA282548141 Enamine

3-[(pyridin-2-ylsulfanyl)methyl]aniline

  • MDL number MFCD11134012
  • Cas Number : 1019522-88-3
  • Molecular Weight : 216.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60575
Availability details for
Quantity
  • Availability
  • Add to Cart