ENA282548144 Enamine

2-[(3-methoxyphenyl)amino]-N-methylacetamide

  • MDL number MFCD11154225
  • Cas Number : 1020940-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56877
Availability details for
Quantity
  • Availability
  • Add to Cart