ENA282548145 Enamine

methyl 4-(chloromethyl)-5-methylfuran-2-carboxylate

  • MDL number MFCD01875643
  • Cas Number : 98490-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60580
Availability details for
Quantity
  • Availability
  • Add to Cart