ENA282548165 Enamine

4-[(2-hydroxyethane)sulfonyl]benzoic acid

  • MDL number MFCD11179735

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60635

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60635
Availability details for
Quantity
  • Availability
  • Add to Cart