ENA282548166 Enamine

6-(2-chloroacetyl)-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD01664324
  • Cas Number : 54903-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-57217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57217
Availability details for
Quantity
  • Availability
  • Add to Cart