ENA282548174 Enamine

3-[5-(2-bromo-5-methoxyphenyl)-1,3,4-oxadiazol-2-yl]propanoic acid

  • MDL number MFCD12871781
  • Cas Number : 1179591-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60644
Availability details for
Quantity
  • Availability
  • Add to Cart