ENA282548177 Enamine

1,3-thiazol-4-ylmethanamine

  • MDL number MFCD06797207
  • Cas Number : 16188-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60647

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60647
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart