ENA282548178 Enamine

9-bromo-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD11206490
  • Cas Number : 1094432-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60648
Availability details for
Quantity
  • Availability
  • Add to Cart