ENA282548189 Enamine

N-butylpiperidine-3-carboxamide hydrochloride

  • MDL number MFCD13562383
  • Cas Number : 937725-10-5
  • Molecular Weight : 220.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60674
Availability details for
Quantity
  • Availability
  • Add to Cart