ENA282548191 Enamine

3,4-dihydro-2H-1-benzopyran-3-amine

  • MDL number MFCD04114513
  • Cas Number : 60575-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57961

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57961
Availability details for
Quantity
  • Availability
  • Add to Cart