ENA282548192 Enamine

3-(propan-2-yl)imidazolidine-2,4-dione

  • MDL number MFCD12193081
  • Cas Number : 63637-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60678
Availability details for
Quantity
  • Availability
  • Add to Cart