ENA282548194 Enamine

3,4-dihydro-2H-1-benzothiopyran-4-ol

  • MDL number MFCD00006883
  • Cas Number : 40316-60-7
  • Molecular Weight : 166.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60680
Availability details for
Quantity
  • Availability
  • Add to Cart