ENA282548206 Enamine

1-[2-(4-ethylpiperazin-1-yl)pyridin-4-yl]methanamine

  • MDL number MFCD09731485
  • Cas Number : 953907-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59010
Availability details for
Quantity
  • Availability
  • Add to Cart