ENA282548222 Enamine

ethyl 6-methyl-4-phenyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD00188414
  • Cas Number : 33458-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60723
Availability details for
Quantity
  • Availability
  • Add to Cart