ENA282548231 Enamine

ethyl(propyl)amine

  • MDL number MFCD00015198
  • Cas Number : 20193-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 4
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35035

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-35035
pictograms GHS02,GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart