ENA282548237 Enamine

(1-bromo-2-phenylethyl)benzene

  • MDL number MFCD02066377
  • Cas Number : 16766-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60746

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60746
Availability details for
Quantity
  • Availability
  • Add to Cart